Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
500 g
Catalog number 010648.22
also known as 010648-22
Price (USD)/ Each
173.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
173.00
Add to cart
Barium nitrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010648.22
Price (USD)/ Each
173.00
-
Add to cart
Chemical Identifiers
CAS10022-31-8
IUPAC Namebarium(2+) dinitrate
Molecular FormulaBaN2O6
InChI KeyIWOUKMZUPDVPGQ-UHFFFAOYSA-N
SMILES[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder
Assay from Suppliers CofA±2.5 % of Theoretical value (Ba)
Total Metal Impurities≤0.001% (10 ppm)
Barium nitrate is used by the oil and gas industries to make drilling muds. It is used to make  paints, bricks, tiles, glass, rubber, and other barium compounds. It also used in is used in signal flares and water softener. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only