Thermo Scientific Chemicals

Bismuth(III) iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010656.09
10 g, Each
Thermo Scientific Chemicals

Bismuth(III) iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010656.09
10 g, Each
Quantity
Catalog number: 010656.09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-64-6
IUPAC Name
bismuth(3+) triiodide
Molecular Formula
BiI3
InChI Key
KOECRLKKXSXCPB-UHFFFAOYSA-K
SMILES
[I-].[I-].[I-].[Bi+3]
Appearance (Color)
Grey to black
Form
Powder and/or granules
Assay (special)
?2.5 % of Theoretical value (Bi)
Total Metal Impurities
≤0.001% (10 ppm)

Description

Polycrystalline films of bismuth triiodide are used as beta direct and digital detectors for medical imaging applications. Bismuth(III) iodide is used as an ice nucleant. It is also used in electron microscopy to reveal synapses. Addition of bismuth triiodide has been shown to improve the glass forming ability and thermal stability of Ge-Te-BiI(3) chalcogenide glasses. Solution-processable BiI3 nanosheets have been shown to exhibit the potential to be used as HTLs (Hole Transport Layer) in conventional OPV (organic photovoltaic) devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text