Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010661.09
also known as 010661-09
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.20 
Save 9.55 (15%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.20 
Save 9.55 (15%)
Add to cart
Cadmium chloride hydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number010661.09
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
62.20 
Save 9.55 (15%)
-
Add to cart
Chemical Identifiers
CAS654054-66-7
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Cadmium chloride hydrate is used in dyeing, printing, textiles and in metal finishing baths. It is also used as a reagent to make photographic paper and carbon paper. Further, it is utilized as an absorbing agent to absorb hydrogen sulfide as well as analytical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only