Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010663.06
5 g, Each
Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010663.06
5 g, Each
Quantity
Catalog number: 010663.06
also known as 010663-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10022-68-1
IUPAC Name
cadmium(2+) bis(nitric acid)
Molecular Formula
CdH2N2O6
InChI Key
IZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
Total Metal Impurities
0.001% max.

Description

It is used for coloring glass and porcelain, and as a flash powder in photography. It is used as a laboratory reagent, in preparation of cadmium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text