Thermo Scientific Chemicals

Calcium carbonate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010679.14
25 g, Each
Thermo Scientific Chemicals

Calcium carbonate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010679.14
25 g, Each
Quantity
Catalog number: 010679.14
also known as 010679-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
471-34-1
IUPAC Name
calcium carbonate
Molecular Formula
CCaO3
InChI Key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
SMILES
[Ca++].[O-]C([O-])=O
Total Metal Impurities
≤0.01% (100 ppm)
Form
Powder
Assay (special)
?2.5 % of Theoretical value (Ca)
Appearance (Color)
White

Description

Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It forms the major component of ceramic tile adhesives. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text