Thermo Scientific Chemicals

Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010683.14
25 g, Each
Thermo Scientific Chemicals

Calcium nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010683.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 010683.14
also known as 010683-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
35054-52-5
IUPAC Name
calcium dinitrate
Molecular Formula
CaN2O6
InChI Key
ZCCIPPOKBCJFDN-UHFFFAOYSA-N
SMILES
[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Metal Impurities
0.005% max.

Description

Calcium nitrate hydrate is used as a nitrate enriching salt. It is also used as a component in fertilizers, waste water treatment to prevent odor and inset accelerating concrete admixtures. Further, it is used for regenerable cold packs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text