Thermo Scientific Chemicals

Calcium oxide, Puratronic™, 99.998% (metals basis, excluding other alkaline earth and alkali metals 130ppm max), Thermo Scientific Chemicals

Catalog number: 010684.09
10 g, Each
Thermo Scientific Chemicals

Calcium oxide, Puratronic™, 99.998% (metals basis, excluding other alkaline earth and alkali metals 130ppm max), Thermo Scientific Chemicals

Catalog number: 010684.09
10 g, Each
Quantity
Catalog number: 010684.09
also known as 010684-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1305-78-8
IUPAC Name
calcium oxidandiide
Molecular Formula
CaO
InChI Key
BRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES
[O--].[Ca++]
Impurity content
other alkaline earth and alkali metals 130ppm max
Total Metal Impurities
0.008% max.

Description

Calcium oxide has wide-ranging utility in industry. It is used in making porcelain, cements, mortars, and glass. It is also employed in flue-gas desulfurization in power plants, purifying sugar, drying the solvents/reagents, preparing bleaching powder, calcium carbide, and calcium cyanamide. It also finds use in water softeners, chemical recovery at Kraft pulp mills, biodiesel production, and in petroleum industry for the detection of traces of water. In agriculture it is used to restore the pH of acidic soils (liming). It is useful as a basic catalyst & acid scavenger in several organic transformations, for example, in Arndt-Eistert Synthesis of alpha-diazoketones, synthesis of heterocycles and oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text