Thermo Scientific Chemicals

Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010686.06
5 g, Each
Thermo Scientific Chemicals

Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010686.06
5 g, Each
Quantity
Catalog number: 010686.06
also known as 010686-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10060-12-5
IUPAC Name
chromium(3+) hexahydrate trichloride
Molecular Formula
Cl3CrH12O6
InChI Key
LJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
Total Metal Impurities
0.002% max.

Description

Chromium(III) chloride hexahydrate is used as an organic synthesis catalyst and as an additive to intravenous solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text