Thermo Scientific Chemicals

Chromium(III) oxide, Puratronic™, 99.97% (metals basis), Thermo Scientific Chemicals

Catalog number: 010688.22
100 g, Each
Thermo Scientific Chemicals

Chromium(III) oxide, Puratronic™, 99.97% (metals basis), Thermo Scientific Chemicals

Catalog number: 010688.22
100 g, Each
Quantity
Catalog number: 010688.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1308-38-9
IUPAC Name
dichromium(3+) trioxidandiide
Molecular Formula
Cr2O3
InChI Key
UOUJSJZBMCDAEU-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Cr+3].[Cr+3]
Total Metal Impurities
0.03% max.

Description

Also known as green compound for buffing and polishing, it can be used as abrasives. Apart from being used as a precursor to the magnetic pigment viz., chromium dioxide, it is used in inks, glasses, and pigments (viridian). They are employed as plating agents and surface treating agents. It is a catalyst in the Phillips process of manufacture of high density polythene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text