Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010694.09
10 g, Each
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010694.09
10 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 010694.09
also known as 010694-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-22-9
IUPAC Name
λ²-cobalt(2+) hexahydrate dinitrate
Molecular Formula
CoH12N2O12
InChI Key
QGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Red to orange
Form
Crystals
Assay (special)
?2.5 % of Theoretical value (Co)
Total Metal Impurities
≤0.001% (10 ppm)

Description

It is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text