Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 010695.18
also known as 010695-18
Price (USD)/ Each
531.65
Online exclusive
591.00 
Save 59.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
531.65
Online exclusive
591.00 
Save 59.35 (10%)
Add to cart
Cobalt(II,III) oxide, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
Catalog number010695.18
Price (USD)/ Each
531.65
Online exclusive
591.00 
Save 59.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1308-06-1
IUPAC Namedicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular FormulaCo3O4
InChI KeyUBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentMay contain up to 10% Cobalt(II) oxide.
CommentNot tested for non-metallic impurities such as sulfur, phosphorus or silicon
Elemental AnalysisCobalt, Co: 73-74%
Total Metal Impurities≤0.0015%
Appearance (Color)Black
View more
It is used in artificial photosynthesis. It is used as a magnetic semiconductor. It is also employed in supercapacitors and lithium-ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only