Thermo Scientific™

Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010696.22
100 g, Each
Thermo Scientific™

Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010696.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 010696.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-24-1
IUPAC Name
λ²-cobalt(2+) heptahydrate sulfate
Molecular Formula
CoH14O11S
InChI Key
MEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
Total Metal Impurities
0.001% max.

Description

Cobalt(II) sulfate heptahydrate is used in the preparation of pigments and other cobalt salts. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text