Thermo Scientific Chemicals

Copper(II) chloride hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010698.22
100 g, Each
Thermo Scientific Chemicals

Copper(II) chloride hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010698.22
100 g, Each
Quantity
Catalog number: 010698.22
also known as 010698-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10125-13-0
IUPAC Name
copper(2+) dihydrate dichloride
Molecular Formula
Cl2CuH4O2
InChI Key
MPTQRFCYZCXJFQ-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Cu++]
Total Metal Impurities
0.001% max.

Description

Copper(II) chloride hydrate promotes the hydrolysis of acetonides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text