Thermo Scientific Chemicals

Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010701.22
100 g, Each
Thermo Scientific Chemicals

Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010701.22
100 g, Each
Quantity
Catalog number: 010701.22
also known as 010701-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23254-43-5
IUPAC Name
copper(2+) sulfate
Molecular Formula
CuO4S
InChI Key
ARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES
[Cu++].[O-]S([O-])(=O)=O
Total Metal Impurities
0.001% max.

Description

It is used as a catalyst in certain organic transformations like synthesis of ynamides. It is used in Fehling's solution and Benedict's solution for testing reducing sugars, in the Biuret reagent for testing proteins, and in combination with potassium permanganate for testing blood for anemia. It is used in the preparation of Cheshunt compound for horticulture. It finds use as a coloring ingredient in artworks, as an oxidant in combination with potassium permanganate for oxidizing primary alcohols, and as a mordant in dyeing. It is used to etch copper or zinc plates for intaglio print making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text