Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010719.06
also known as 010719-06
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
21.30 
Save 2.65 (12%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
21.30 
Save 2.65 (12%)
Add to cart
Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number010719.06
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
21.30 
Save 2.65 (12%)
-
Add to cart
Chemical Identifiers
CAS6080-56-4
IUPAC Nameλ²-lead(2+) diacetate trihydrate
Molecular FormulaC4H12O7Pb
InChI KeyMCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only