Thermo Scientific Chemicals

Lead(II) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010722.14
25 g, Each
Thermo Scientific Chemicals

Lead(II) chloride, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010722.14
25 g, Each
Quantity
Catalog number: 010722.14
also known as 010722-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7758-95-4
IUPAC Name
λ²-lead(2+) dichloride
Molecular Formula
Cl2Pb
InChI Key
HWSZZLVAJGOAAY-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Pb++]
Appearance (Color)
White
Total Metal Impurities
≤0.001% (10 ppm)
Form
Crystalline

Description

Lead chloride is used in the synthesis of lead titanate and barium lead titanate ceramics; employed in the production of infrared transmitting glass and ornamental glass (aurene glass); useful as an electrode in geophysical applications and in solar cells. Analytical reagent, preparation of lead salts, as solder and flux. Pattinson's white lead, pigment in white paint, is the basic lead chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text