Thermo Scientific Chemicals

Lead(II) fluoride, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010723.14
25 g, Each
Thermo Scientific Chemicals

Lead(II) fluoride, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010723.14
25 g, Each
Quantity
Catalog number: 010723.14
also known as 010723-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-46-2
IUPAC Name
λ²-lead(2+) difluoride
Molecular Formula
F2Pb
InChI Key
FPHIOHCCQGUGKU-UHFFFAOYSA-L
SMILES
[F-].[F-].[Pb++]
Total Metal Impurities
0.003% max.

Description

Lead(II) fluoride is used in fuses, glass coatings to reflect infrared rays and phosphors for television screens and in low melting glasses. Further, it serves as a catalyst for the preparation of picoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text