Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010727.18
also known as 010727-18
Price (USD)/ Each
155.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
183.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
155.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
183.00 
Save 27.35 (15%)
Add to cart
Lead(II) nitrate, White powder, Puratronic™, 99.999% (Metals basis), Thermo Scientific Chemicals
Catalog number010727.18
Price (USD)/ Each
155.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
183.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10099-74-8
IUPAC Nameλ²-lead(2+) dinitrate
Molecular FormulaN2O6Pb
InChI KeyRLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Employed in industrial applications such as heat stabilization in nylon and polyesters, coatings of photothermographic paper, gold cyanidation and pyrotechnics. Finds application in supramolecular chemistry. Strong oxidizing agent, and an alternative to the Sommelet reaction for the oxidation of benzylic halides to aldehydes. Used in the preparation of isothiocyanates from dithiocarbamates. Scavenger of bromide in nucleophilic substitution reactions. Manufacture of dinitrogen tetroxide in indurtrial scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only