Lead(II) oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Lead(II) oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific™

Lead(II) oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010728.22
also known as 010728-22
Price (USD)/ Each
476.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
561.00 
Save 84.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
476.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
561.00 
Save 84.35 (15%)
Add to cart
Lead(II) oxide, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals
Catalog number010728.22
Price (USD)/ Each
476.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
561.00 
Save 84.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1317-36-8
IUPAC Nameplumbanone
Molecular FormulaOPb
InChI KeyYEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILESO=[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0005% max.
It is used in the manufacture of glass, ceramics and computer components. Addition of lead oxide increases the refractive index, electrical resistivity, Curie temperature, and the ability of the glass to absorb X-rays. It is also added to decrease the viscosity of glass. It is employed in the vulcanization of rubber, lead acid batteries, and in the production of certain pigments and paints. In cathode ray tube glass lead oxide is used to block X-ray emission. It acts as an efficient reagent in oxidation reactions as well as condensation reactions of hydroxy ketones with diamines to form quinoxaline derivatives with high yield.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only