Thermo Scientific Chemicals

Lead(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010729.09
10 g, Each
Thermo Scientific Chemicals

Lead(IV) oxide, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010729.09
10 g, Each
Quantity
Catalog number: 010729.09
also known as 010729-09
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1309-60-0
IUPAC Name
plumbanedione
Molecular Formula
O2Pb
InChI Key
YADSGOSSYOOKMP-UHFFFAOYSA-N
SMILES
O=[Pb]=O
Total Metal Impurities
0.005% max.

Description

It is employed in manufacture of matches and electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text