Thermo Scientific Chemicals

Lithium bromide hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010733.14
25 g, Each
Thermo Scientific Chemicals

Lithium bromide hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010733.14
25 g, Each
Quantity
Catalog number: 010733.14
also known as 010733-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85017-82-9
IUPAC Name
lithium(1+) bromide
Molecular Formula
BrLi
InChI Key
AMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES
[Li+].[Br-]
Total Metal Impurities
0.005% max.

Description

Lithium bromide is used as a medium for air-conditioning, and a desiccant industrial drying system. It finds use as a sedative and hypnotic. Further it is employed in manufacturing pharmaceuticals, in alkylation processes, and as brazing and welding fluxes. In addition, lithium bromide-water solution is widely used in absorption refrigeration systems, absorption heat pumps and absorption heat transformers. l-Proline lithium bromide monohydrate (LPLBM) is a promising semi-organic nonlinear optical material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text