Thermo Scientific Chemicals

Lithium carbonate, Puratronic™, 99.998% (metals basis excluding Ca), Ca LT 20ppm, Thermo Scientific Chemicals

Catalog number: 010734.18
50 g, Each
Thermo Scientific Chemicals

Lithium carbonate, Puratronic™, 99.998% (metals basis excluding Ca), Ca LT 20ppm, Thermo Scientific Chemicals

Catalog number: 010734.18
50 g, Each
Quantity
Catalog number: 010734.18
also known as 010734-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
554-13-2
IUPAC Name
dilithium(1+) carbonate
Molecular Formula
CLi2O3
InChI Key
XGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]C([O-])=O
Total Metal Impurities
≤0.002% (20 ppm)
Appearance (Color)
White
Form
Powder
Assay (special)
?2.5 % of Theoretical value (Li)

Description

Stainless Steel powder is employed in pharmaceuticals, marine applications, architectural applications, medical implants, including pins, screws and orthopaedic implants like total hip and knee replacements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text