Thermo Scientific Chemicals

Lithium chloride hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 010735.14
25 g, Each
Thermo Scientific Chemicals

Lithium chloride hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 010735.14
25 g, Each
Quantity
Catalog number: 010735.14
also known as 010735-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85144-11-2
IUPAC Name
lithium(1+) chloride
Molecular Formula
ClLi
InChI Key
KWGKDLIKAYFUFQ-UHFFFAOYSA-M
SMILES
[Li+].[Cl-]
Total Metal Impurities
0.004% max.

Description

Lithium chloride is widely employed in lithium metal production. It is used as a brazing flux for aluminum in automobile parts. Its use can also be seen as a desiccant for drying air streams. In biochemical applications, lithium chloride is used to precipitate RNA from cellular extracts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text