Thermo Scientific Chemicals

Lithium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals, Puratronic™

Catalog number: 010736.09
10 g, Each
Thermo Scientific Chemicals

Lithium fluoride, 99.99% (metals basis), Thermo Scientific Chemicals, Puratronic™

Catalog number: 010736.09
10 g, Each
Quantity
Catalog number: 010736.09
also known as 010736-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-24-4
IUPAC Name
lithium(1+) fluoride
Molecular Formula
FLi
InChI Key
PQXKHYXIUOZZFA-UHFFFAOYSA-M
SMILES
[Li+].[F-]
Appearance (Color)
White
Total Metal Impurities
≤0.01% (100 ppm)
Form
Powder

Description

Used in specialized UV optics. Also used as a crystal in X-ray spectrometry. Widely employed in PLED (Polymer Light Emitting Diodes) and OLED (Organic Light Emitting Diodes) as a coupling layer to enhance electron injection. Found in liquid-fluoride nuclear reactors. Used to record ionizing radiation in thermoluminescent dosimeters. Component of molten salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text