Lithium metaborate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Lithium metaborate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium metaborate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 010739.22
also known as 010739-22
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
Add to cart
Lithium metaborate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
Catalog number010739.22
Price (USD)/ Each
271.65
Online exclusive
302.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13453-69-5
IUPAC Nametetraboron heptahydrate dilithium
Molecular FormulaB4H14Li2O7
InChI KeyDPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.003% max.
Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only