Thermo Scientific Chemicals

Lithium phosphate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010743.14
25 g, Each
Thermo Scientific Chemicals

Lithium phosphate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 010743.14
25 g, Each
Quantity
Catalog number: 010743.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-52-3
IUPAC Name
trilithium(1+) phosphate
Molecular Formula
Li3O4P
InChI Key
TWQULNDIKKJZPH-UHFFFAOYSA-K
SMILES
[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O
Total Metal Impurities
0.01% max.

Description

Lithium phosphate batteries batteries are now widely used as a staple for cell phones, laptops and some other gadgets. Used in low-expansion porcelain enamel glazes and polymer intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text