Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals
Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010753.22
also known as 010753-22
Price (USD)/ Each
504.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
593.00 
Save 88.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
504.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
593.00 
Save 88.35 (15%)
Add to cart
Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm, Thermo Scientific Chemicals
Catalog number010753.22
Price (USD)/ Each
504.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
593.00 
Save 88.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.02% max., Nb 50ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only