Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 010755.09
also known as 010755-09
Price (USD)/ Each
37.65
Online exclusive
41.90 
Save 4.25 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.90 
Save 4.25 (10%)
Add to cart
Sulfur pieces, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010755.09
Price (USD)/ Each
37.65
Online exclusive
41.90 
Save 4.25 (10%)
-
Add to cart
Chemical Identifiers
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only