Aluminum wire, 1.5mm (0.059in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Aluminum wire, 1.5mm (0.059in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum wire, 1.5mm (0.059in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Form: Approx. 4.8g/m
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 m
1 m
Catalog number 010765.G5
also known as 010765-G5
Price (USD)/ Each
144.00
-
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (USD)/ Each
144.00
Add to cart
Aluminum wire, 1.5mm (0.059in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010765.G5
Price (USD)/ Each
144.00
-
Add to cart
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.999% (metals basis)
Appearance (Color)Silvery white
FormWire, 1.5mm (0.059in) diameter, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only