Tin Foil, 0.025mm (0.001 in.) thick, hard, 99.9% (Metals basis), Thermo Scientific Chemicals
Tin Foil, 0.025mm (0.001 in.) thick, hard, 99.9% (Metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin Foil, 0.025mm (0.001 in.) thick, hard, 99.9% (Metals basis), Thermo Scientific Chemicals

Form: Approx. 4.2g/15 x 15cm; Temper: as rolled
Have Questions?
Change viewbuttonViewtableView
Quantity:
15 x 15 cm
15 x 75 cm
Catalog number 010789.VA
also known as 010789-VA
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Quantity:
15 x 15 cm
Request bulk or custom format
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
Add to cart
Tin Foil, 0.025mm (0.001 in.) thick, hard, 99.9% (Metals basis), Thermo Scientific Chemicals
Catalog number010789.VA
Price (USD)/ Each
144.65
Online exclusive
160.00 
Save 15.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-31-5
IUPAC Nametin
Molecular FormulaSn
InChI KeyATJFFYVFTNAWJD-UHFFFAOYSA-N
SMILES[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only