Thermo Scientific Chemicals

Magnesium carbonate hydroxide hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 010796.14
25 g, Each
Thermo Scientific Chemicals

Magnesium carbonate hydroxide hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 010796.14
25 g, Each
Quantity
Catalog number: 010796.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39409-82-0
IUPAC Name
pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular Formula
C4H10Mg5O18
InChI Key
YMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILES
O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Total Metal Impurities
0.004% max.

Description

Magnesium carbonate hydroxide hydrate is used as drier, color ficatives, anticaking agent, carrier, swelling agent, acidity regulator. Magnesium carbonate is the production of magnesium oxide by calcining. Magnesite and dolomite minerals are used to produce refractory bricks. MgCO3 is also used in flooring, fireproofing, fire extinguishing compositions, cosmetics, dusting powder, and toothpaste. Other applications are as filler material, smoke suppressant in plastics, a reinforcing agent in neoprene rubber, a drying agent, a laxative to loosen the bowels, and color retention in foods. In addition, high purity magnesium carbonate is used as antacid and as an additive in table salt to keep it free flowing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text