Thermo Scientific Chemicals

Magnesium chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010797.22
100 g, Each
Thermo Scientific Chemicals

Magnesium chloride hexahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010797.22
100 g, Each
Quantity
Catalog number: 010797.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-18-6
IUPAC Name
magnesium(2+) hexahydrate dichloride
Molecular Formula
Cl2H12MgO6
InChI Key
DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
Total Metal Impurities
0.001% max.

Description

Utilized in the polymerase chain reactions (PCR). Used for dust control, ice control, and road stabilization. Employed in a variety of applications such as fertilizer, mineral supplement for animals, waste water treatment, artificial seawater, textiles, paper, fireproofing agents, cements, and fire extinguishers. Used in several medicines and topical or skin related applications. Acts as a coagulant in the preparation of tofu from soy milk. Acts as an anaesthetic for cephalopods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text