Thermo Scientific Chemicals

Magnesium nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010799.06
5 g, Each
Thermo Scientific Chemicals

Magnesium nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010799.06
5 g, Each
Quantity
Catalog number: 010799.06
also known as 010799-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-60-3
IUPAC Name
magnesium(2+) dinitrate
Molecular Formula
MgN2O6
InChI Key
YIXJRHPUWRPCBB-UHFFFAOYSA-N
SMILES
[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Total Metal Impurities
0.001% max.

Description

It is used in printing, chemical, agriculture and ceramics industries. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text