Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 010802.06
also known as 010802-06
Price (USD)/ Each
67.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
67.50
Add to cart
Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010802.06
Price (USD)/ Each
67.50
-
Add to cart
Chemical Identifiers
CAS6156-78-1
IUPAC Namemanganese(2+) diacetate tetrahydrate
Molecular FormulaC4H14MnO8
InChI KeyCESXSDZNZGSWSP-UHFFFAOYSA-L
SMILESO.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Used as a desiccant, fertilizer and a catalyst. Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst. Used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only