Thermo Scientific Chemicals

Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010802.14
25 g, Each
Thermo Scientific Chemicals

Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010802.14
25 g, Each
Quantity
Catalog number: 010802.14
also known as 010802-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6156-78-1
IUPAC Name
manganese(2+) diacetate tetrahydrate
Molecular Formula
C4H14MnO8
InChI Key
CESXSDZNZGSWSP-UHFFFAOYSA-L
SMILES
O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
Total Metal Impurities
0.001% max.

Description

Used as a desiccant, fertilizer and a catalyst. Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst. Used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text