Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number 010806.18
also known as 010806-18
Price (USD)/ Each
289.65
Online exclusive
322.00 
Save 32.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
289.65
Online exclusive
322.00 
Save 32.35 (10%)
Add to cart
Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number010806.18
Price (USD)/ Each
289.65
Online exclusive
322.00 
Save 32.35 (10%)
-
Add to cart
Chemical Identifiers
CAS15710-66-4
IUPAC Namemanganese(2+) dinitrate
Molecular FormulaMnN2O6
InChI KeyMIVBAHRSNUNMPP-UHFFFAOYSA-N
SMILES[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Raw material for electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. Also useful precursors to the oxides of manganese.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only