Thermo Scientific Chemicals

Manganese(II) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010807.14
25 g, Each
Thermo Scientific Chemicals

Manganese(II) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010807.14
25 g, Each
Quantity
Catalog number: 010807.14
also known as 010807-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15244-36-7
IUPAC Name
manganese(2+) hydrate sulfate
Molecular Formula
H2MnO5S
InChI Key
ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES
O.[Mn++].[O-]S([O-])(=O)=O
Total Metal Impurities
0.001% max.

Description

Used in feed additive, nutrient, firming agent and flavor enhancer. It is also used as a fermentation aid in the processing of beer and malt beverages.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text