Thermo Scientific Chemicals

Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max, Thermo Scientific Chemicals

Catalog number: 010812.22
100 g, Each
Thermo Scientific Chemicals

Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max, Thermo Scientific Chemicals

Catalog number: 010812.22
100 g, Each
Quantity
Catalog number: 010812.22
also known as 010812-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-27-5
IUPAC Name
trioxomolybdenum
Molecular Formula
MoO3
InChI Key
JKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILES
O=[Mo](=O)=O
Total Metal Impurities
0.002% max.
Impurity content
W 300ppm max.

Description

Its chief application is as an oxidation catalyst and as a raw material for the production of molybdenum metal. It is a Precursor to LAMOX fast ion conductors and superconductors. Molybdenum trioxide has also been suggested as a potential anti-microbial agent, e.g., in polymers. In contact with water, it forms H+ ions that can kill bacteria effectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text