Thermo Scientific Chemicals

Nickel(II) oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010819.06
5 g, Each
Thermo Scientific Chemicals

Nickel(II) oxide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 010819.06
5 g, Each
Quantity
Catalog number: 010819.06
also known as 010819-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-99-1
IUPAC Name
oxonickel
Molecular Formula
NiO
InChI Key
GNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILES
O=[Ni]
Form
Crystalline powder or granules
Appearance (Color)
Grey-green to dark-green
Assay (special)
±2.5 % of Theoretical value (Ni)
Total Metal Impurities
≤0.002% (20 ppm)

Description

Nickel (II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text