Zinc powder, average 4-7 micron, 97.5% (metals basis), Thermo Scientific Chemicals
Zinc powder, average 4-7 micron, 97.5% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc powder, average 4-7 micron, 97.5% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
5 x 1 kg
Catalog number 010835.A1
Price (USD)/ Each
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
Total Metal Impurities≤2.5%
Particle Size5-8 micron average
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only