Thermo Scientific Chemicals

Potassium sulfate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010845.14
25 g, Each
Thermo Scientific Chemicals

Potassium sulfate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010845.14
25 g, Each
Quantity
Catalog number: 010845.14
also known as 010845-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-80-5
IUPAC Name
dipotassium sulfate
Molecular Formula
K2O4S
InChI Key
OTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S([O-])(=O)=O
Total Metal Impurities
0.003% max.

Description

Potassium sulfate principal use is as a fertilizer. It is also used occasionally in the production of glass and as a flash reducer in artillery propellant charges. It reduces muzzle flash, flareback and blast overpressure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text