Rubidium nitrate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Rubidium nitrate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Thermo Scientific™

Rubidium nitrate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 010852.18
also known as 010852-18
Price (USD)/ Each
623.65
Online exclusive
693.00 
Save 69.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
623.65
Online exclusive
693.00 
Save 69.35 (10%)
Add to cart
Rubidium nitrate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals
Catalog number010852.18
Price (USD)/ Each
623.65
Online exclusive
693.00 
Save 69.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13126-12-0
IUPAC Namerubidium(1+) nitrate
Molecular FormulaNO3Rb
InChI KeyRTHYXYOJKHGZJT-UHFFFAOYSA-N
SMILES[Rb+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.025% max.
Rubidium nitrate is used in infrared radiation producing pyrotechnic compositions. It acts as a colorant and an oxidizer especially in decoys and illumination flares. Further, it is employed as a starting material in the preparation of other rubidium compounds and rubidium metal, which is utilized for the preparation of catalysts and in scintillation counters. In addition to this, it is used in fireworks to produce a red-violet color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only