Thermo Scientific Chemicals

Silicon(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010856.22
100 g, Each
Thermo Scientific Chemicals

Silicon(IV) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 010856.22
100 g, Each
Quantity
Catalog number: 010856.22
also known as 010856-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Total Metal Impurities
0.001% max.

Description

Silica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text