Thermo Scientific Chemicals

Silver chloride, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010857.14
25 g, Each
Thermo Scientific Chemicals

Silver chloride, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010857.14
25 g, Each
Quantity
Catalog number: 010857.14
also known as 010857-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-90-6
IUPAC Name
silver(1+) chloride
Molecular Formula
AgCl
InChI Key
HKZLPVFGJNLROG-UHFFFAOYSA-M
SMILES
[Cl-].[Ag+]
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay from Suppliers CofA
≥99.997 (metals basis)
Total Metal Impurities
≤0.003%

Description

Employed in Silver plating. Owing to its characteristic of reversible reduction to silver metal, it is used in photochromic lenses. Used as a cathode in sea water activated batteries. In electrochemistry, silver chloride electrode is used for potentiometric measurements. It serves as an antidote for mercury poisoning, and eliminates mercury from body. It is used in glass manufacturing industry. It is useful in the production of bandages, wound healing products and inglaze lustre, personal deodorant products, as well as for long term preservation of drinking water in water tanks; its pharmaceutical composition finds use as an antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text