Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 010870.14
also known as 010870-14
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
Add to cart
Sodium dihydrogen phosphate monohydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number010870.14
Price (USD)/ Each
129.65
Online exclusive
144.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10049-21-5
IUPAC Namesodium phosphoric acid hydrate
Molecular FormulaH5NaO5P
InChI KeyBBMHARZCALWXSL-UHFFFAOYSA-N
SMILESO.[Na+].OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Sodium dihydrogen phosphate monohydrate is used as a food additive. It finds use as mordant in dyeing. It is also used in the manufacture of enamels, chemicals, pharmaceuticals, ceramics, detergents, and boiler compounds. In addition, it is used as a fireproofing agent, thickening agent, emulsifier, and as a reagent and buffer in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only