Sodium iodide hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Sodium iodide hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium iodide hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 010871.14
also known as 010871-14
Price (USD)/ Each
122.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
122.00
Add to cart
Sodium iodide hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Catalog number010871.14
Price (USD)/ Each
122.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialSodium iodide hydrate
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
Recommended StorageAmbient temperatures
View more
Sodium Iodide is used in the synthesis of dye-sensitized solar cells. Also used in the preparation of propargylamines as inhibitors of lysine specific. Demethylase. Used in photography and as analytical reagents. They are used in the measurement of the energy of gamma rays, by measuring the amplitude of pulses of light generated by electrons which are excited by the gamma rays and in the Finkelstein reaction via an SN2 pathway, detection of radiation, and in the conversion of an alkyl chloride into an alkyl iodide. Sodium Iodide is used in PCR and to produce chemokines, particularly CCL2, CXCL8, and CXC14 by stimulation of thyroid follicular cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only