Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 010875.14
also known as 010875-14
Price (USD)/ Each
74.65
Online exclusive
83.00 
Save 8.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
74.65
Online exclusive
83.00 
Save 8.35 (10%)
Add to cart
Sodium tetraborate, anhydrous, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
Catalog number010875.14
Price (USD)/ Each
74.65
Online exclusive
83.00 
Save 8.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1330-43-4
IUPAC Namedisodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular FormulaB4Na2O7
InChI KeyNXSVMNYRVBQCMO-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.002% max.
Sodium tetraborate is used as a co-catalyst for (2,2,6,6-tetramethylpiperidin-1-yl)oxyl and sodium hypochlorite oxidation of alcohols in greener and non-chlorinated solvents. It acts as a reagent for the decomposition of oxides and silicates by fusion. It is used to prepare buffers, which is used in gel electrophoresis of DNA. It acts as a source of borate, micronutrient fertilizer, preservative in taxidermy, additive in ceramic slips and glazes. It is also used as a precursor for sodium perborate monohydrate and boric acid. It finds application as fluoride detoxification, neutron absorber in nuclear reactors and to make indelible ink for dip pens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only