Thermo Scientific Chemicals

Strontium Fluoride, 99.99% (Metal Basis), Thermo Scientific Chemicals, Puratronic™

Catalog number: 010878.22
100 g, Each
Thermo Scientific Chemicals

Strontium Fluoride, 99.99% (Metal Basis), Thermo Scientific Chemicals, Puratronic™

Catalog number: 010878.22
100 g, Each
Quantity
Catalog number: 010878.22
also known as 010878-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-48-4
IUPAC Name
strontium(2+) difluoride
Molecular Formula
F2Sr
InChI Key
FVRNDBHWWSPNOM-UHFFFAOYSA-L
SMILES
[F-].[F-].[Sr++]
Total Metal Impurities
0.01% max.

Description

Strontium fluoride is used as an optical coating on lenses, and as a thermoluminescent dosimeter crystal. It also finds use as a water insoluble strontium source for use in oxygen-sensitive applications such as metal production. It serves both as a source of strontium and fluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text