Tin(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals
Tin(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tin(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 010891.14
also known as 010891-14
Price (USD)/ Each
460.65
Online exclusive
512.00 
Save 51.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
460.65
Online exclusive
512.00 
Save 51.35 (10%)
Add to cart
Tin(IV) oxide, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals
Catalog number010891.14
Price (USD)/ Each
460.65
Online exclusive
512.00 
Save 51.35 (10%)
-
Add to cart
Chemical Identifiers
CAS18282-10-5
IUPAC Namestannanedione
Molecular FormulaO2Sn
InChI KeyXOLBLPGZBRYERU-UHFFFAOYSA-N
SMILESO=[Sn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.004% max.
Tin(IV) oxide is used as a pigment in the ceramic industry and as a polishing powder. It finds use in sensors of combustible gases. It is also used as a catalyst along with vanadium oxide for the oxidation of aromatic compounds to prepare carboxylic acids and acid anhydrides. It is used in sensors for detection of trace levels of carbon monoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only