Thermo Scientific Chemicals

Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010894.06
5 g, Each
Thermo Scientific Chemicals

Tin(II) chloride hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 010894.06
5 g, Each
Quantity
Catalog number: 010894.06
also known as 010894-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1370709-86-6
Total Metal Impurities
0.005% max.

Description

Tin(II) chloride is widely used as a reducing agent in organic synthesis, for example conversion of quinones into hydroquinones, nitroaromatics into aromatic amines, and in Stephen reduction. It can be used to reduce certain metal salts into metals, tin plating, analytical testing of mercury ions, formation of silver mirrors and metal-metal bonds. It is also used for tin-plating, as a mordant in textile dyeing and as a catalyst in the production of polylactic acid. Further it is used as a food additive, wherein it serves as an anti-oxidant and a color retention agent. It can be used in the condensation of aryl aldehydes with cyclohexan-1,3-diones to synthesize xanthenes. It catalyzes direct conversion of aldehydes into beta-keto esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text